siRNA Details
| Gene Symbol: Pdgfc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019971.2 | NP_064355.1 | 4 | 1704 | ||
| GenBank mRNA | AF117608.1 | - | 4 | 880 | ||
| AF266467.1 | - | 4 | 1704 | |||
| AK033734.1 | - | 4 | 1433 | |||
| AK042767.1 | - | 4 | 1451 | |||
| AK052947.1 | - | 4 | 956 | |||
| AK081347.1 | - | 5 | 756 | |||
| BC029099.1 | - | 5 | 1007 | |||
| BC037696.1 | - | 4 | 891 | 4008749 | ||

Pdgfc
Genomic Map