siRNA Details
| Gene Symbol: Praf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138602.4 | NP_613068.1 | 3 | 490 | ||
| GenBank mRNA | AF229636.1 | - | 3 | 477 | ||
| AK003344.1 | - | 3 | 491 | |||
| BC064757.1 | - | 3 | 515 | 30056020 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Praf2| Gene Symbol: Praf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138602.4 | NP_613068.1 | 3 | 490 | ||
| GenBank mRNA | AF229636.1 | - | 3 | 477 | ||
| AK003344.1 | - | 3 | 491 | |||
| BC064757.1 | - | 3 | 515 | 30056020 | ||