siRNA Details
| Gene Symbol: Sfmbt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001166531.1 | NP_001160003.1 | 9 | 1152 | ||
| NM_001166532.1 | NP_001160004.1 | 8 | 1246 | |||
| NM_019460.2 | NP_062333.1 | 8 | 1103 | |||
| GenBank mRNA | AB032165.1 | - | 7 | 948 | ||
| AK142330.1 | - | 8 | 1130 | |||
| AK154826.1 | - | 8 | 1248 | |||
| AK160573.1 | - | 9 | 1152 | |||
| BC018217.1 | - | 8 | 1114 | |||
| BC036972.1 | - | 8 | 1173 | |||
| BC066188.1 | - | 8 | 1056 | 30476111 | ||

Sfmbt1
Genomic Map