siRNA Details
| Gene Symbol: Zfp108 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018791.2 | NP_061261.2 | 4 | 561 | ||
| GenBank mRNA | AF167315.1 | - | 4 | 532 | ||
| AK032355.1 | - | 4 | 562 | |||
| BC100339.1 | - | 4 | 506 | 30939834 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp108| Gene Symbol: Zfp108 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018791.2 | NP_061261.2 | 4 | 561 | ||
| GenBank mRNA | AF167315.1 | - | 4 | 532 | ||
| AK032355.1 | - | 4 | 562 | |||
| BC100339.1 | - | 4 | 506 | 30939834 | ||