siRNA Details
| Gene Symbol: Hs3st3b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018805.2 | NP_061275.2 | 2 | 2128 | ||
| GenBank mRNA | AK085242.1 | - | 2 | 2136 | ||
| AK171996.1 | - | 2 | 2122 | |||
| AK172015.1 | - | 2 | 2127 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hs3st3b1| Gene Symbol: Hs3st3b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018805.2 | NP_061275.2 | 2 | 2128 | ||
| GenBank mRNA | AK085242.1 | - | 2 | 2136 | ||
| AK171996.1 | - | 2 | 2122 | |||
| AK172015.1 | - | 2 | 2127 | |||