siRNA Details
| Gene Symbol: Plxnc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018797.2 | NP_061267.1 | 7 | 1736 | ||
| GenBank mRNA | AF190578.1 | - | 7 | 1736 | ||
| AK084603.1 | - | 7 | 1376 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Plxnc1| Gene Symbol: Plxnc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018797.2 | NP_061267.1 | 7 | 1736 | ||
| GenBank mRNA | AF190578.1 | - | 7 | 1736 | ||
| AK084603.1 | - | 7 | 1376 | |||