siRNA Details
| Gene Symbol: Ctps2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001168568.1 | NP_001162040.1 | 16 | 1762 | ||
| NM_001168569.1 | NP_001162041.1 | 16 | 1819 | |||
| NM_018737.5 | NP_061207.1 | 16 | 1894 | |||
| GenBank mRNA | AK032018.1 | - | 16 | 1621 | ||
| AK052854.1 | - | 16 | 1810 | |||
| AK084595.1 | - | 16 | 1691 | |||
| AK085381.1 | - | 16 | 1670 | |||
| AK150966.1 | - | 16 | 1761 | |||
| AK151597.1 | - | 16 | 1761 | |||
| BC003257.1 | - | 16 | 1602 | 3500778 | ||
| EF410142.1 | - | 493 | ||||
| U49385.1 | - | 16 | 1784 | |||

Ctps2
Genomic Map