siRNA Details
| Gene Symbol: Slc13a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019481.2 | NP_062354.2 | 3 | 307 | ||
| GenBank mRNA | AF199365.1 | - | 3 | 307 | ||
| AF199366.1 | - | 3 | 307 | |||
| AK019876.1 | - | 3 | 306 | |||
| AK128899.1 | - | 3 | 307 | |||
| BC040789.1 | - | 3 | 281 | |||
| BC049981.1 | - | 3 | 289 | 5065026 | ||

Slc13a1
Genomic Map