siRNA Details
| Gene Symbol: Impa1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018864.5 | NP_061352.2 | 3 | 247 | ||
| GenBank mRNA | AF042730.1 | - | 3 | 192 | ||
| AK011770.1 | - | 3 | 185 | |||
| AK013728.1 | - | 2 | 183 | |||
| AK078406.1 | - | 3 | 196 | |||
| AK083238.1 | - | 3 | 181 | |||
| AK145472.1 | - | 184 | ||||
| AK147778.1 | - | 3 | 207 | |||
| AK152774.1 | - | 3 | 198 | |||
| AK153457.1 | - | 3 | 214 | |||
| AK160203.1 | - | 3 | 185 | |||
| AK160467.1 | - | 3 | 179 | |||
| AK165977.1 | - | 3 | 189 | |||
| AK168457.1 | - | 3 | 222 | |||
| BC049080.1 | - | 3 | 188 | 6419835 | ||
| BC055722.1 | - | 2 | 142 | 6413389 | ||
| BY283562.1 | - | 3 | 248 | |||

Impa1
Genomic Map