siRNA Details
| Gene Symbol: Paxip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018878.3 | NP_061366.2 | 13, 14 | 2712 | ||
| GenBank mRNA | AF104261.1 | - | 13, 14 | 2712 | ||
| AK144606.1 | - | 13, 14 | 2574 | |||
| AK148141.1 | - | 13, 14 | 2583 | |||
| BC066014.1 | - | 13, 14 | 2501 | 5704566 | ||

Paxip1
Genomic Map