siRNA Details
| Gene Symbol: Pdzrn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018884.2 | NP_061372.2 | 10 | 2392 | ||
| GenBank mRNA | AF127084.1 | - | 10 | 2392 | ||
| AF127085.1 | - | 11 | 2236 | |||
| AK143226.1 | - | 10 | 1906 | |||
| AK173098.1 | - | 10 | 2351 | |||
| BC010329.1 | - | 6 | 1212 | 3600603 | ||
| HM560981.1 | - | 9 | 2284 | |||

Pdzrn3
Genomic Map