siRNA Details
| Gene Symbol: Cxcl13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018866.2 | NP_061354.1 | 2 | 125 | ||
| GenBank mRNA | AF030636.1 | - | 2 | 121 | ||
| AF044196.1 | - | 2 | 125 | |||
| AK156025.1 | - | 2 | 139 | |||
| AK160241.1 | - | 2 | 141 | |||
| BB612247.1 | - | 136 | ||||
| BC012965.1 | - | 2 | 124 | 4217539 | ||

Cxcl13
Genomic Map