siRNA Details
| Gene Symbol: Fmo2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018881.3 | NP_061369.2 | 2 | 148 | ||
| GenBank mRNA | AF184981.1 | - | 1 | 213 | ||
| AK009224.1 | - | 2 | 183 | |||
| BC031415.1 | - | 2 | 148 | 3990305 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fmo2| Gene Symbol: Fmo2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018881.3 | NP_061369.2 | 2 | 148 | ||
| GenBank mRNA | AF184981.1 | - | 1 | 213 | ||
| AK009224.1 | - | 2 | 183 | |||
| BC031415.1 | - | 2 | 148 | 3990305 | ||