siRNA Details
| Gene Symbol: Smad9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019483.5 | NP_062356.3 | 6 | 1549 | ||
| GenBank mRNA | AF175408.2 | - | 5 | 1227 | ||
| AK137411.1 | - | 6 | 1550 | |||
| AY145520.1 | - | 5 | 1382 | |||
| BC119141.1 | - | 5 | 1266 | 8733890 | ||

Smad9
Genomic Map