siRNA Details
| Gene Symbol: Cyp39a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001285947.1 | NP_001272876.1 | 2 | 469 | ||
| NM_001285948.1 | NP_001272877.1 | 2 | 469 | |||
| NM_018887.4 | NP_061375.1 | 2 | 469 | |||
| GenBank mRNA | AF237981.1 | - | 2 | 318 | ||
| AK031733.1 | - | 2 | 417 | |||
| BC021418.1 | - | 2 | 340 | 5025117 | ||
| BC037663.1 | - | 2 | 318 | 5352740 | ||

Cyp39a1
Genomic Map