siRNA Details
| Gene Symbol: Cyp39a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001285947.1 | NP_001272876.1 | 9 | 1275 | ||
| NM_018887.4 | NP_061375.1 | 10 | 1425 | |||
| GenBank mRNA | AF237981.1 | - | 10 | 1274 | ||
| AK031733.1 | - | 10 | 1373 | |||
| BC037663.1 | - | 9 | 1124 | 5352740 | ||

Cyp39a1
Genomic Map