siRNA Details
| Gene Symbol: Cxcl11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019494.1 | NP_062367.1 | 2 | 241 | ||
| GenBank mRNA | AF136449.1 | - | 2 | 241 | ||
| AF178672.1 | - | 2 | 241 | |||
| AF179872.1 | - | 2 | 241 | |||
| AK020750.1 | - | 2 | 260 | |||
| AK040051.1 | - | 2 | 201 | |||
| AK050012.1 | - | 2448 | ||||
| BC025903.1 | - | 2 | 229 | 5035983 | ||

Cxcl11
Genomic Map