siRNA Details
| Gene Symbol: KRT32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002278.3 | NP_002269.3 | 4 | 916 | ||
| GenBank mRNA | X81419.1 | - | 4 | 755 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

KRT32| Gene Symbol: KRT32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002278.3 | NP_002269.3 | 4 | 916 | ||
| GenBank mRNA | X81419.1 | - | 4 | 755 | ||