siRNA Details
| Gene Symbol: Pdss1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019501.3 | NP_062374.2 | 5 | 620 | ||
| GenBank mRNA | AB210841.1 | - | 5 | 540 | ||
| AF118855.1 | - | 3 | 315 | |||
| AK012307.1 | - | 5 | 548 | |||
| BC026820.1 | - | 5 | 522 | 4039464 | ||
| BC107273.1 | - | 5 | 435 | |||
| BC107274.1 | - | 5 | 439 | |||

Pdss1
Genomic Map