siRNA Details
| Gene Symbol: Dgke | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019505.3 | NP_062378.1 | 2 | 931 | ||
| GenBank mRNA | AF136744.1 | - | 2 | 568 | ||
| AK044596.1 | - | 1667 | ||||
| AK049335.1 | - | 2 | 731 | |||
| AK147841.1 | - | 2 | 932 | |||
| BB635763.1 | - | 2 | 335 | |||
| BC055867.1 | - | 2 | 864 | |||

Dgke
Genomic Map