siRNA Details
| Gene Symbol: Astn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019514.3 | NP_062387.2 | 17, 18 | 3213 | ||
| NM_207109.2 | NP_996992.1 | 18, 19 | 3369 | |||
| GenBank mRNA | AF116573.1 | - | 15, 16 | 2381 | ||
| AK041160.1 | - | 19, 20 | 3273 | |||

Astn2
Genomic Map