siRNA Details
| Gene Symbol: Bace2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019517.4 | NP_062390.3 | 7 | 1146 | ||
| GenBank mRNA | AF051150.1 | - | 3 | 295 | ||
| AF216310.1 | - | 7 | 1097 | |||
| AK052309.1 | - | 7 | 1133 | |||
| AK078770.1 | - | 7 | 1113 | |||
| AK165036.1 | - | 7 | 1082 | |||
| AK185329.1 | - | 4 | 386 | |||
| BC120773.1 | - | 7 | 1112 | 40129696 | ||
| BC131947.1 | - | 7 | 1112 | 40130224 | ||

Bace2
Genomic Map