siRNA Details
| Gene Symbol: Rbm38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019547.2 | NP_062420.2 | 4 | 697 | ||
| GenBank mRNA | AI573584.1 | - | 4 | 716 | ||
| AK052886.1 | - | 2 | 482 | |||
| AK146026.1 | - | 4 | 671 | |||
| AK162141.1 | - | 4 | 670 | |||
| AK172625.1 | - | 4 | 668 | |||
| BC006687.1 | - | 3 | 1206 | |||
| BC085307.1 | - | 4 | 641 | 6396103 | ||

Rbm38
Genomic Map