siRNA Details
| Gene Symbol: Rbm38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019547.2 | NP_062420.2 | 4 | 805 | ||
| GenBank mRNA | AK052886.1 | - | 2 | 590 | ||
| AK146026.1 | - | 4 | 779 | |||
| AK162141.1 | - | 4 | 778 | |||
| BC006687.1 | - | 3 | 1314 | |||
| BC085307.1 | - | 4 | 749 | 6396103 | ||

Rbm38
Genomic Map