siRNA Details
| Gene Symbol: Prpf40a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018785.2 | NP_061255.1 | 10 | 749 | ||
| GenBank mRNA | AF135439.1 | - | 10 | 749 | ||
| AK087963.1 | - | 10 | 666 | |||
| BC003318.1 | - | 10 | 776 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prpf40a| Gene Symbol: Prpf40a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018785.2 | NP_061255.1 | 10 | 749 | ||
| GenBank mRNA | AF135439.1 | - | 10 | 749 | ||
| AK087963.1 | - | 10 | 666 | |||
| BC003318.1 | - | 10 | 776 | |||