siRNA Details
| Gene Symbol: Uchl5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159866.1 | NP_001153338.1 | 7, 8 | 751 | ||
| NM_019562.2 | NP_062508.2 | 7, 8 | 751 | |||
| GenBank mRNA | AF148447.1 | - | 7, 8 | 694 | ||
| AF175903.1 | - | 7, 8 | 703 | |||
| AK007860.1 | - | 6, 7 | 565 | |||
| AK011117.1 | - | 7, 8 | 718 | |||
| AK017925.1 | - | 7, 8 | 749 | |||
| BC006891.1 | - | 7, 8 | 704 | 2650799 | ||

Uchl5
Genomic Map