siRNA Details
| Gene Symbol: Uchl5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159866.1 | NP_001153338.1 | 9 | 925 | ||
| NM_019562.2 | NP_062508.2 | 9 | 928 | |||
| GenBank mRNA | AF148447.1 | - | 9 | 871 | ||
| AF175903.1 | - | 9 | 877 | |||
| AK007860.1 | - | 8 | 739 | |||
| AK011117.1 | - | 9 | 895 | |||
| AK017925.1 | - | 9 | 923 | |||
| BC006891.1 | - | 9 | 878 | 2650799 | ||

Uchl5
Genomic Map