siRNA Details
| Gene Symbol: Htra1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019564.3 | NP_062510.2 | 3 | 776 | ||
| GenBank mRNA | AF172994.1 | - | 3 | 752 | ||
| AF179369.1 | - | 3 | 747 | |||
| AK090320.1 | - | 3 | 491 | |||
| AK128916.1 | - | 3 | 749 | |||
| AK132270.1 | - | 3 | 749 | |||
| AK167667.1 | - | 3 | 342 | |||
| BC013516.1 | - | 3 | 746 | 4235912 | ||
| CB193609.1 | - | 3 | 777 | |||

Htra1
Genomic Map