siRNA Details
| Gene Symbol: Patz1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001253690.1 | NP_001240619.1 | 2 | 1483 | ||
| NM_019574.3 | NP_062520.1 | 2 | 1483 | |||
| GenBank mRNA | AB029397.1 | - | 2 | 1483 | ||
| AK018372.1 | - | 2 | 237 | |||
| BC026673.1 | - | 2 | 154 | |||

Patz1
Genomic Map