siRNA Details
| Gene Symbol: Tspan5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019571.5 | NP_062517.1 | 5 | 861 | ||
| GenBank mRNA | AF121344.1 | - | 5 | 876 | ||
| AK013350.1 | - | 5 | 826 | |||
| AK015705.1 | - | 5 | 583 | |||
| AK033543.1 | - | 5 | 560 | |||
| AK187703.1 | - | 4 | 375 | |||
| BC058695.1 | - | 5 | 832 | 6813872 | ||

Tspan5
Genomic Map