siRNA Details
| Gene Symbol: KRT84 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033045.3 | NP_149034.2 | 1 | 129 | ||
| XM_011538335.2 | XP_011536637.1 | 2 | 244 | |||
| GenBank mRNA | AJ628420.1 | - | 1 | 75 | ||
| AK314030.1 | - | 1 | 148 | |||
| BC069647.1 | - | 1 | 129 | 7262449 | ||

KRT84
Genomic Map