siRNA Details
| Gene Symbol: Styx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019637.3 | NP_062611.2 | 3 | 398 | ||
| GenBank mRNA | AK002822.1 | - | 340 | |||
| AK030122.1 | - | 3 | 398 | |||
| AK042741.1 | - | 3 | 362 | |||
| AK046970.1 | - | 3 | 375 | |||
| AK154136.1 | - | 3 | 493 | |||
| AK166043.1 | - | 3 | 381 | |||
| BC038608.1 | - | 3 | 348 | 5012303 | ||
| BC085089.1 | - | 3 | 345 | 6492329 | ||
| BC096674.1 | - | 3 | 406 | 5144003 | ||
| U34973.1 | - | 3 | 366 | |||

Styx
Genomic Map