siRNA Details
| Gene Symbol: Arl6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019665.3 | NP_062639.3 | 6 | 878 | ||
| GenBank mRNA | AB232697.1 | - | 5 | 441 | ||
| AF031903.1 | - | 6 | 596 | |||
| AK003778.1 | - | 6 | 706 | |||
| AK152980.1 | - | 6 | 682 | |||
| AK158832.1 | - | 6 | 931 | |||
| AK160666.1 | - | 6 | 697 | |||
| BC018497.1 | - | 6 | 698 | 3484868 | ||

Arl6
Genomic Map