siRNA Details
| Gene Symbol: Pitpnb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001301643.1 | NP_001288572.1 | 9 | 750 | ||
| NM_001301644.1 | NP_001288573.1 | 9 | 719 | |||
| NM_001301666.1 | NP_001288595.1 | 8 | 666 | |||
| NM_019640.5 | NP_062614.1 | 9 | 750 | |||
| GenBank mRNA | AK028235.1 | - | 9 | 685 | ||
| BC034676.1 | - | 9 | 640 | 4481685 | ||
| U46934.1 | - | 9 | 652 | |||

Pitpnb
Genomic Map