siRNA Details
| Gene Symbol: Metap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019648.3 | NP_062622.1 | 12 | 1921 | ||
| GenBank mRNA | AB003144.1 | - | 11 | 1232 | ||
| AF434712.1 | - | 11 | 1311 | |||
| AK053691.1 | - | 3 | 1302 | |||
| AK076804.1 | - | 11 | 1264 | |||
| AK090323.1 | - | 11 | 1250 | |||
| AK147097.1 | - | 11 | 1356 | |||
| AK159650.1 | - | 11 | 1335 | |||
| AK163318.1 | - | 11 | 1338 | |||
| AK165301.1 | - | 12 | 1923 | |||
| AK165611.1 | - | 2 | 99 | |||
| AK165845.1 | - | 11 | 1247 | |||
| AK194694.1 | - | 1 | 57 | |||
| BC002213.1 | - | 11 | 1333 | 3490341 | ||
| BC010263.1 | - | 11 | 1305 | |||
| BC092052.1 | - | 11 | 1298 | 3670685 | ||

Metap2
Genomic Map