siRNA Details
| Gene Symbol: Zfp113 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019747.4 | NP_062721.2 | 5 | 1335 | ||
| GenBank mRNA | AK076349.1 | - | 5 | 1330 | ||
| AK220556.1 | - | 5 | 1335 | |||
| BC052453.1 | - | 5 | 1279 | 6410632 | ||
| BC054525.1 | - | 5 | 1279 | 6410632 | ||
| BC056445.1 | - | 4 | 1200 | 5703904 | ||
| BC056991.1 | - | 5 | 1281 | |||

Zfp113
Genomic Map