siRNA Details
| Gene Symbol: Dnajb5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019874.3 | NP_063927.1 | 1 | 281 | ||
| GenBank mRNA | AF088983.1 | - | 2 | 328 | ||
| AF321322.1 | - | 2 | 321 | |||
| AK158535.1 | - | 2 | 373 | |||
| AK158972.1 | - | 2 | 581 | |||
| BC027782.1 | - | 1 | 3024 | |||
| BC048902.1 | - | 2 | 566 | 6396037 | ||
| BC057087.1 | - | 1 | 281 | 5695243 | ||

Dnajb5
Genomic Map