siRNA Details
| Gene Symbol: Stam2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019667.2 | NP_062641.1 | 8 | 815 | ||
| GenBank mRNA | AB012611.1 | - | 8 | 830 | ||
| AK004604.2 | - | 8 | 815 | |||
| AK035789.1 | - | 868 | ||||
| AK044230.1 | - | 7 | 744 | |||
| AK078244.1 | - | 8 | 847 | |||
| AK159699.1 | - | 8 | 844 | |||
| AK168729.1 | - | 8 | 847 | |||
| BC013818.1 | - | 8 | 851 | 3671426 | ||

Stam2
Genomic Map