siRNA Details
| Gene Symbol: Eif3c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146200.1 | NP_666312.1 | 17, 18 | 2388 | ||
| GenBank mRNA | AK088076.1 | - | 17, 18 | 2431 | ||
| AK152693.1 | - | 17, 18 | 2406 | |||
| AK160078.1 | - | 17, 18 | 2408 | |||
| AK168160.1 | - | 17, 18 | 2392 | |||
| AK168174.1 | - | 17, 18 | 2392 | |||
| AK212354.1 | - | 1, 2 | 149 | |||
| BC024855.1 | - | 17, 18 | 2388 | 5359060 | ||
| BC025032.1 | - | 17, 18 | 2409 | 4947435 | ||
| BC080763.1 | - | 4, 5 | 803 | |||
| U67328.1 | - | 6, 7 | 1160 | |||

Eif3c
Genomic Map