siRNA Details
Gene Symbol: Gltp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019821.2 | NP_062795.2 | 2 | 224 | ||
GenBank mRNA | AF209703.1 | - | 2 | 150 | ||
AK157313.1 | - | 2 | 211 | |||
BC016584.1 | - | 2 | 224 | 3157800 | ||
BC053729.1 | - | 2 | 167 | 30065192 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
