siRNA Details
| Gene Symbol: Pus1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025561.3 | NP_001020732.1 | 6 | 1502 | ||
| NM_001025562.2 | NP_001020733.1 | 5 | 1145 | |||
| NM_019700.4 | NP_062674.2 | 5 | 1448 | |||
| GenBank mRNA | AB041563.1 | - | 6 | 1504 | ||
| AF116237.1 | - | 4 | 1105 | |||
| AK089258.1 | - | 5 | 1448 | |||
| AK138570.1 | - | 3 | 1023 | |||
| BC021446.1 | - | 5 | 1324 | 5038201 | ||
| BC034359.1 | - | 5 | 1419 | 3595162 | ||

Pus1
Genomic Map