siRNA Details
| Gene Symbol: Clcnkb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019701.2 | NP_062675.2 | 16 | 1774 | ||
| GenBank mRNA | AA240610.1 | - | 2 | 176 | ||
| AF124847.1 | - | 16 | 1767 | |||
| AK146876.1 | - | 15 | 1837 | |||
| BC019983.1 | - | 16 | 1800 | 4188062 | ||

Clcnkb
Genomic Map