siRNA Details
| Gene Symbol: Cpb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019775.3 | NP_062749.2 | 4, 5 | 445 | ||
| GenBank mRNA | AA048136.1 | - | 4, 5 | 445 | ||
| AB021968.1 | - | 4, 5 | 393 | |||
| AF164524.1 | - | 4, 5 | 460 | |||
| AF186188.1 | - | 4, 5 | 396 | |||
| AK004045.1 | - | 4, 5 | 418 | |||
| BC089577.1 | - | 4, 5 | 417 | 6766672 | ||

Cpb2
Genomic Map