siRNA Details
| Gene Symbol: LAG3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002286.5 | NP_002277.4 | 5 | 1299 | ||
| XM_011520956.1 | XP_011519258.1 | 5 | 1320 | |||
| GenBank mRNA | A21353.1 | - | 5 | 1180 | ||
| AK292104.1 | - | 5 | 1273 | |||
| BC052589.1 | - | 5 | 1284 | 6301931 | ||
| X51985.3 | - | 5 | 1180 | |||

LAG3
Genomic Map