siRNA Details
| Gene Symbol: Trip4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170907.1 | NP_001164378.1 | 1, 2 | 239 | ||
| NM_019797.4 | NP_062771.2 | 4, 5 | 1055 | |||
| GenBank mRNA | AF197574.1 | - | 1, 2 | 114 | ||
| AF539614.1 | - | 4, 5 | 938 | |||
| AK039024.1 | - | 1, 2 | 124 | |||
| AK078393.1 | - | 1, 2 | 116 | |||
| AK146298.1 | - | 1, 2 | 122 | |||
| AK150752.1 | - | 1, 2 | 116 | |||
| BC021316.1 | - | 1, 2 | 127 | 5027899 | ||

Trip4
Genomic Map