siRNA Details
| Gene Symbol: Nudt3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001291046.1 | NP_001277975.1 | 5 | 634 | ||
| NM_019837.2 | NP_062811.1 | 5 | 626 | |||
| GenBank mRNA | AF264064.1 | - | 5 | 625 | ||
| AK079658.1 | - | 3 | 121 | |||
| BC016534.1 | - | 6 | 532 | 2615904 | ||
| BC046805.1 | - | 5 | 627 | 6829170 | ||
| BU919402.1 | - | 5 | 458 | |||

Nudt3
Genomic Map