siRNA Details
| Gene Symbol: Cbln3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019820.3 | NP_062794.1 | 1 | 309 | ||
| GenBank mRNA | AF218379.1 | - | 1 | 285 | ||
| AK081085.1 | - | 1 | 284 | |||
| BC065108.1 | - | 1 | 309 | 30536445 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cbln3| Gene Symbol: Cbln3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019820.3 | NP_062794.1 | 1 | 309 | ||
| GenBank mRNA | AF218379.1 | - | 1 | 285 | ||
| AK081085.1 | - | 1 | 284 | |||
| BC065108.1 | - | 1 | 309 | 30536445 | ||