siRNA Details
| Gene Symbol: Diap3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019670.1 | NP_062644.1 | 23, 24 | 2809 | ||
| GenBank mRNA | AF094519.1 | - | 23, 24 | 2809 | ||
| AK138694.1 | - | 23, 24 | 2839 | |||
| AK161737.1 | - | 22, 23 | 2827 | |||
| AK166882.1 | - | 23, 24 | 2865 | |||
| AK220493.1 | - | 23, 24 | 2873 | |||
| BC028920.1 | - | 16, 17 | 2110 | 4008236 | ||
| BC085191.1 | - | 23, 24 | 2919 | 30693873 | ||
| BC086779.1 | - | 23, 24 | 2884 | |||

Diap3
Genomic Map