siRNA Details
| Gene Symbol: Diap3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019670.1 | NP_062644.1 | 12 | 1283 | ||
| GenBank mRNA | AF094519.1 | - | 12 | 1283 | ||
| AK137073.1 | - | 11 | 1372 | |||
| AK138694.1 | - | 12 | 1313 | |||
| AK161737.1 | - | 11 | 1301 | |||
| AK166882.1 | - | 12 | 1339 | |||
| AK220493.1 | - | 12 | 1347 | |||
| BC003250.1 | - | 11 | 1317 | |||
| BC028920.1 | - | 5 | 584 | 4008236 | ||
| BC085191.1 | - | 12 | 1393 | 30693873 | ||
| BC086779.1 | - | 12 | 1358 | |||

Diap3
Genomic Map