siRNA Details
| Gene Symbol: Pdcd10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019745.4 | NP_062719.2 | 3, 4 | 406 | ||
| GenBank mRNA | AF159368.1 | - | 3, 4 | 309 | ||
| AK005594.1 | - | 3, 4 | 335 | |||
| AK075926.1 | - | 3, 4 | 354 | |||
| AK077308.1 | - | 3, 4 | 383 | |||
| AK137198.1 | - | 3, 4 | 376 | |||
| AK149182.1 | - | 3, 4 | 312 | |||
| AK160234.1 | - | 3, 4 | 356 | |||
| AK168857.1 | - | 3, 4 | 340 | |||
| AK169689.1 | - | 3, 4 | 340 | |||
| BC019650.1 | - | 3, 4 | 346 | 4022513 | ||
| BC086778.1 | - | 3, 4 | 325 | 5689088 | ||

Pdcd10
Genomic Map